4-Phenylbutyric acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Phenylbutanoic acid
M-Clarity™ quality level = MQ200
P21005-100G
P21005-25G
SIALP21005-100GEA
4580
CZK
SIALP21005-100G
SIALP21005-25G
4-Phenylbutyric acid, Sigma-Aldrich®
4-Phenylbutyric acid
Vzorec:
C₁₀H₁₂O₂ MW: 164,2 g/mol Bod varu: 290…294 °C (1013 hPa) Bod tání: 48…51 °C |
MDL Number:
MFCD00004403 CAS číslo: 1821-12-1 EINECS: 217-341-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...