Tisk…

3-Phenylpropionic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 3-Phenylpropanoic acid, Hydrocinnamic acid

A14908.22 A14908.36
A14908.22EA 1810 CZK
A14908.22 A14908.36
3-Phenylpropionic acid ≥99%
3-Phenylpropionic acid

Purity: 99% CAS No.: 501-52-0 Molecular Formula: C6H5(CH2)2CO2H Formula Weight: 150.18 Flash Point: 110°(230°F) Melting Point: 47-50° Boiling Point: 280° Beilstein Reference: 907515 Merck Reference: 14,4784 MDL No.: MFCD00002771

Vzorec: C₉H₁₀O₂
Bod varu: 280 °C
Bod tání: 47…50 °C
Hustota: 1,071
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002771
CAS číslo: 501-52-0
EINECS: 207-924-5
Merck Index: 14,4784

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR