(2-Chloro-4-fluorophenyl)acetic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B22872.06
B22872.14
B22872.06EA
3000
CZK
B22872.06
B22872.14
(2-Chloro-4-fluorophenyl)acetic acid ≥97%
(2-Chloro-4-fluorophenyl)acetic acid
Purity: 97%
CAS No.: 177985-32-9
Molecular Formula: C8H6ClFO2
Formula Weight: 188.59
Melting Point: 102-105°
MDL No.: MFCD00236028
Vzorec:
(C₆H₃ClF)CH₂COOH Bod varu: 289 °C (1013 hPa) Bod tání: 102…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236028 CAS číslo: 177985-32-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...