4-Nitrophenyl chloroformate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A18742.06
A18742.14
A18742.22
AB124931.0025
A18742.06EA
1020
CZK
A18742.06
A18742.14
A18742.22
ABCRAB124931.0025
4-Nitrophenyl chloroformate ≥97%
4-Nitrophenyl chloroformate
Purity: 97%
CAS No.: 7693-46-1
Molecular Formula: C7H4ClNO4
Formula Weight: 201.57
Storage Temperature: 2°C to 8°C
Flash Point:
Vzorec:
C₇H₄ClNO₄ Bod varu: 159…162 °C (19 mmHg) Bod tání: 75…82 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007321 CAS číslo: 7693-46-1 EINECS: 231-706-9 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...