Tisk…

(4-Bromophenyl)acetic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Bromophenylacetic acid, 2-(4-bromophenyl)acetic acid

M-Clarity™ quality level = MQ200

138673-100G 138673-25G
SIAL138673-100GEA 4890 CZK
SIAL138673-100G SIAL138673-25G
(4-Bromophenyl)acetic acid, Sigma-Aldrich®
(4-Bromophenyl)acetic acid
Vzorec: (C₆H₄Br)CH₂COOH
MW: 215.05
Bod varu: 326
Bod tání: 94…98 °C
MDL Number: MFCD00004342
CAS číslo: 1878-68-8
EINECS: 217-523-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR