(4-Bromophenyl)acetic acid ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-Bromophenylacetic acid, 2-(4-bromophenyl)acetic acid
A15325.14
A15325.22
A15325.14EA
2320
CZK
A15325.14
A15325.22
(4-Bromophenyl)acetic acid ≥99%
(4-Bromophenyl)acetic acid
Purity: 99%
CAS No.: 1878-68-8
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 115-119°
Beilstein Reference: 972146
Vzorec:
(C₆H₄Br)CH₂COOH MW: 215.05 Bod varu: 326 Bod tání: 115…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004342 CAS číslo: 1878-68-8 EINECS: 217-523-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...