(2-Bromophenyl)acetic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-(2-Bromophenyl)acetic acid
M-Clarity™ quality level = MQ200
260061-25G
260061-5G
SIAL260061-25GEA
3580
CZK
SIAL260061-25G
SIAL260061-5G
(2-Bromophenyl)acetic acid, Sigma-Aldrich®
(2-Bromophenyl)acetic acid
Vzorec:
(C₆H₄Br)CH₂COOH MW: 215.05 Bod varu: 308 Bod tání: 107.5 |
MDL Number:
MFCD00004314 CAS číslo: 18698-97-0 EINECS: 242-509-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...