(2-Bromophenyl)acetic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-(2-Bromophenyl)acetic acid
A16900.06
A16900.14
A16900.22
A16900.06EA
1320
CZK
A16900.06
A16900.14
A16900.22
(2-Bromophenyl)acetic acid ≥98%
(2-Bromophenyl)acetic acid
Purity: 98+%
CAS No.: 18698-97-0
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 105-106°
Beilstein Reference: 2250655
Vzorec:
(C₆H₄Br)CH₂COOH MW: 215,05 g/mol Bod varu: 308 °C (1013 hPa) Bod tání: 105…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004314 CAS číslo: 18698-97-0 EINECS: 242-509-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...