3-(Indol-3-yl)propionic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Indole-3-propionic acid, 3-Indolepropionic acid
M-Clarity™ quality level = MQ200
220027-10G
220027-1G
SIAL220027-10GEA
1770
CZK
SIAL220027-10G
SIAL220027-1G
3-(Indol-3-yl)propionic acid, Sigma-Aldrich®
3-(Indol-3-yl)propionic acid
Vzorec:
C₁₁H₁₁NO₂ MW: 189,21 g/mol Bod tání: 129…132 °C Hustota: 1,356 g/cm³ (20 °C) |
MDL Number:
MFCD00005660 CAS číslo: 830-96-6 EINECS: 212-600-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...