Tisk…

Indol-3-acetic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: IAA, 1H-Indole-3-acetic acid, Heteroauxin, 3-Indoleacetic acid

M-Clarity™ quality level = MQ200

I3750-100G-A I3750-25G-A I3750-5G-A
SIALI3750-100G-AEA 9040 CZK
SIALI3750-100G-A SIALI3750-25G-A SIALI3750-5G-A
Indol-3-acetic acid, Sigma-Aldrich®
Indol-3-acetic acid
Vzorec: C₁₀H₉NO₂
MW: 175,19 g/mol
Bod varu: 120…125 °C (0,05 mmHg)
Bod tání: 167…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005636
CAS číslo: 87-51-4
EINECS: 201-748-2
Merck Index: 13,04986

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR