Indol-3-acetic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
IAA, 1H-Indole-3-acetic acid, Heteroauxin, 3-Indoleacetic acid
M-Clarity™ quality level = MQ200
I2886-100G
I2886-25G
I2886-5G
SIALI2886-100GEA
12300
CZK
SIALI2886-100G
SIALI2886-25G
SIALI2886-5G
Indol-3-acetic acid, Sigma-Aldrich®
Indol-3-acetic acid
Vzorec:
C₁₀H₉NO₂ MW: 175,19 g/mol Bod varu: 120…125 °C (0,05 mmHg) Bod tání: 167…170 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005636 CAS číslo: 87-51-4 EINECS: 201-748-2 Merck Index: 13,04986 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...