3-Cyclopentylpropionic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
3-Cyclopentylpropanoic acid, 3-Cylcopentylpropanoic acid
M-Clarity™ quality level = MQ100
C115908-100ML
C115908-25ML
SIALC115908-100MLEA
5390
CZK
SIALC115908-100ML
SIALC115908-25ML
3-Cyclopentylpropionic acid, Sigma-Aldrich®
3-Cyclopentylpropionic acid
Vzorec:
C₈H₁₄O₂ Bod varu: 130 °C (1013 hPa) Hustota: 1,01 g/cm³ (17 °C) |
MDL Number:
MFCD00001392 CAS číslo: 140-77-2 EINECS: 205-433-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...