Tisk…

(3,5-Bis(trifluoromethyl)phenyl)acetic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

A18922.03 A18922.06
A18922.03EA 821 CZK
A18922.03 A18922.06
(3,5-Bis(trifluoromethyl)phenyl)acetic acid ≥97%
(3,5-Bis(trifluoromethyl)phenyl)acetic acid

Purity: 97% CAS No.: 85068-33-3 Molecular Formula: C10H6F6O2 Formula Weight: 272.15 Melting Point: 121-123° Beilstein Reference: 6813447

Vzorec: (C₆H₃(CF₃)₂)CH₂COOH
MW: 272.15
Bod varu: 276
Bod tání: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00009908
CAS číslo: 85068-33-3
EINECS: 285-295-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR