Tisk…

2,2-Dimethyl-1,3-propanediol ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Neopentyl Glycol, 2,2-Dimethylpropane-1,3-diol

A10340.0E A10340.30 A10340.36
A10340.0EEA 2770 CZK
A10340.0E A10340.30 A10340.36
2,2-Dimethyl-1,3-propanediol ≥99%
2,2-Dimethyl-1,3-propanediol

Purity: 99% CAS No.: 126-30-7 Molecular Formula: C5H12O2 Formula Weight: 104.15 Flash Point: 103°(217°F) Odour: Mild Boiling Point: 210-212° Beilstein Reference: 605291 Merck Reference: 14,6458 MDL No.: MFCD00004685

Vzorec: C₅H₁₂O₂
MW: 104,15 g/mol
Bod varu: 210…212 °C
Bod tání: 125…130 °C
Hustota: 1,042
Bod vzplanutí: 103 °C (217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004685
CAS číslo: 126-30-7
EINECS: 204-781-0
Merck Index: 14,6458

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR