4-Methyl-1-pentanol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Methylpentan-1-ol
M-Clarity™ quality level = MQ100
M66951-25G
M66951-5G
SIALM66951-25GEA
10140
CZK
SIALM66951-25G
SIALM66951-5G
4-Methyl-1-pentanol, Sigma-Aldrich®
4-Methyl-1-pentanol
Vzorec:
C₆H₁₄O Bod varu: 152 °C (1013 hPa) Bod tání: –70 °C Hustota: 0,84 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002962 CAS číslo: 626-89-1 EINECS: 210-969-3 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...