Tisk…

terc-Butanol ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol

L12338.AK L12338.AU L12338.0F
L12338.AKEA 838 CZK
L12338.AK L12338.AU L12338.0F
terc-Butanol ≥99%
terc-Butanol

Purity: 99% CAS No.: 75-65-0 Molecular Formula: (CH3)3COH Formula Weight: 74.12 Flash Point: 11°(52°F) Refractive Index: 1.3870 Melting Point: 25° Boiling Point: 83° Beilstein Reference: 906698 Merck Reference: 14,1542 UN No.: 1120 MDL No.: MFCD00004464

Vzorec: (CH₃)₃COH
Bod varu: 83 °C
Bod tání: 25 °C
Hustota: 0,786
Bod vzplanutí: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS číslo: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3,II
Merck Index: 14,1542
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR