2-Methyl-2-pentanol ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
A19090.06
A19090.14
A19090.22
A19090.06EA
1370
CZK
A19090.06
A19090.14
A19090.22
2-Methyl-2-pentanol ≥99%
2-Methyl-2-pentanol
Purity: 99%
CAS No.: 590-36-3
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 30°(86°F)
Refractive Index: 1.4100
Odour: Alcohol-like
Melting Point: -103°
Boiling Point: 120-122°
Beilstein Reference: 1718951
UN No.: 2560
MDL No.: MFCD00004485
Vzorec:
C₆H₁₄O MW: 102,18 g/mol Bod varu: 120…122 °C Bod tání: –103 °C Hustota: 0,810 Bod vzplanutí: 30 °C (86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004485 CAS číslo: 590-36-3 EINECS: 209-681-0 UN: 2560 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...