Izobutanol 99+% ACS
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol
36643.AP
36643.K2
36643.M1
36643.APEA
1840
CZK
36643.AP
36643.K2
36643.M1
Izobutanol 99+% ACS
Izobutanol
Purity: 99+%
CAS No.: 78-83-1
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: 28°(82°F)
Physical Form: Liquid
Density: 0.802
Refractive Index: 1.3960
Appearance: Colourless
Odour: Sweet
pH: 7
Melting Point: -108°
Boiling Point: 107-108°
Beilstein Reference: 1730878
Merck Reference: 14,5131
UN No.: 1212
MDL No.: MFCD00004740
Vzorec:
(CH₃)₂CHCH₂OH MW: 74.12 Bod varu: 107…108 °C Bod tání: –108 °C Hustota: 0.802 Bod vzplanutí: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004740 CAS číslo: 78-83-1 EINECS: 201-148-0 UN: 1212 ADR: 3 III Merck Index: 14,5131 |