2,4-Dimethyl-3-pentanol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,4-Dimethylpentan-3-ol
M-Clarity™ quality level = MQ100
150045-100G
150045-25G
SIAL150045-100GEA
2690
CZK
SIAL150045-100G
SIAL150045-25G
2,4-Dimethyl-3-pentanol, Sigma-Aldrich®
2,4-Dimethyl-3-pentanol
Vzorec:
C₇H₁₆O MW: 116,2 g/mol Bod varu: 139 °C (1013 hPa) Bod tání: –70 °C Hustota: 0,8445 g/cm³ (0 °C) |
MDL Number:
MFCD00008921 CAS číslo: 600-36-2 EINECS: 209-993-7 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...