Tisk…

2-Ethylhexyl nitrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

293784-1L 293784-2.5L
SIAL293784-1LEA 2380 CZK
SIAL293784-1L SIAL293784-2.5L
2-Ethylhexyl nitrate, Sigma-Aldrich®
2-Ethylhexyl nitrate
Vzorec: C₈H₁₇NO₃
CAS číslo: 27247-96-7
EINECS: 248-363-6
UN: 3139
ADR: 5.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR