Diisobutylamine 99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L15495.AE
L15495.AP
L15495.AU
L15495.AEEA
951
CZK
L15495.AE
L15495.AP
L15495.AU
Diisobutylamine 99%
Diisobutylamine
Purity: 99%
CAS No.: 110-96-3
Molecular Formula: C8H19N
Formula Weight: 129.25
Flash Point: 30°(86°F)
Refractive Index: 1.4090
Melting Point: -77°
Boiling Point: 137-139°
Beilstein Reference: 1209251
UN No.: 2361
MDL No.: MFCD00008930
Vzorec:
C₈H₁₉N Bod varu: 137…139 °C Bod tání: –77 °C Hustota: 0,744 Bod vzplanutí: 30 °C (86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008930 CAS číslo: 110-96-3 EINECS: 203-819-3 UN: 2361 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...