Tisk…

Bis(2-ethylhexyl)amine 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L13894.AE L13894.AP
L13894.AEEA 1060 CZK
L13894.AE L13894.AP
Bis(2-ethylhexyl)amine 99%
Bis(2-ethylhexyl)amine

Purity: 99% CAS No.: 106-20-7 Molecular Formula: C16H35N Formula Weight: 241.46 Flash Point: 121°(250°F) Refractive Index: 1.4420 Melting Point: -60° Boiling Point: 281° Beilstein Reference: 1748342 UN No.: 2922 MDL No.: MFCD00009489

Vzorec: C₁₆H₃₅N
Bod varu: 281 °C
Bod tání: < –60 °C
Hustota: 0,805
Bod vzplanutí: 121 °C (250 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009489
CAS číslo: 106-20-7
EINECS: 203-372-4
UN: 2922
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR