Isopentylamine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-Amino-3-methylbutane, Isopentyl amine, Isoamylamine, 3-Methylbutylamine, Leucamine
M-Clarity™ quality level = MQ300
W321907-100G
W321907-1KG
W321907-4KG
W321907-SAMPLE
SIALW321907-100GEA
1020
CZK
SIALW321907-100G
SIALW321907-1KG
SIALW321907-4KG
SIALW321907-SAMPLE
Isopentylamine, Sigma-Aldrich®
Isopentylamine
Vzorec:
C₅H₁₃N Bod varu: 97 °C (1013 hPa) Bod tání: –68 °C Hustota: 0,75 g/cm³ (20 °C) Bod vzplanutí: –1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008203 CAS číslo: 107-85-7 EINECS: 203-526-0 UN: 1106 ADR: 3,II Merck Index: 13,05128 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...