2-Ethylhexylamin 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-Ethyl-1-hexylamine
A10372.0F
A10372.AE
A10372.0FEA
5630
CZK
A10372.0F
A10372.AE
2-Ethylhexylamin 98%
2-Ethylhexylamin
Purity: 98%
CAS No.: 104-75-6
Molecular Formula: C8H19N
Formula Weight: 129.25
Flash Point: 50°(122°F)
Refractive Index: 1.4300
Odour: Amine-like
Melting Point: -76°
Boiling Point: 169°
Beilstein Reference: 1209249
UN No.: 2276
MDL No.: MFCD00008148
Vzorec:
C₈H₁₉N MW: 129.25 Bod varu: 169 °C Bod tání: –76 °C Hustota: 0.7882 Bod vzplanutí: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008148 CAS číslo: 104-75-6 EINECS: 203-233-8 UN: 2276 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...