3,3-Dimethylbutyric acid ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3,3-Dimethylbutanoic acid, tert-Butylacetic acid
B21660.14
B21660.22
B21660.14EA
999
CZK
B21660.14
B21660.22
3,3-Dimethylbutyric acid ≥98%
3,3-Dimethylbutyric acid
Purity: 98%
CAS No.: 1070-83-3
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 88° (190°F)
Refractive Index: 1.4110
Odour: Stench
Melting Point: 6°
Boiling Point: 185-190°
Beilstein Reference: 1740427
Merck Reference: 14,1538
UN No.: 3265
MDL No.: MFCD00002715
Vzorec:
C₆H₁₂O₂ MW: 116,16 g/mol Bod varu: 185…190 °C Bod tání: 6 °C Hustota: 0,912 Bod vzplanutí: 88° (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002715 CAS číslo: 1070-83-3 EINECS: 213-982-2 UN: 3265 ADR: 8,III Merck Index: 14,1538 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...