Tisk…

(±)-2-Methylcaproic acid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: 2-MethylhexanoicAcid, 2-Methylhexanoic acid, (±)-2-Methylhexanoic acid

M-Clarity™ quality level = MQ300

W319104-100G-K W319104-1KG-K
SIALW319104-100G-KEA 5100 CZK
SIALW319104-100G-K SIALW319104-1KG-K
(±)-2-Methylcaproic acid, Sigma-Aldrich®
(±)-2-Methylcaproic acid
Vzorec: C₇H₁₄O₂
MW: 130.19
Bod varu: 209
Hustota: 0.929
Storage Temperature: Ambient
MDL Number: MFCD00002674
CAS číslo: 4536-23-6
EINECS: 224-883-9
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR