Tisk…

Pivalic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,2-Dimethylpropionic acid, Trimethylacetic acid

M-Clarity™ quality level = MQ200

T71803-100ML T71803-500ML T71803-5ML
SIALT71803-100MLEA 1240 CZK
SIALT71803-100ML SIALT71803-500ML SIALT71803-5ML
Pivalic acid, Sigma-Aldrich®
Pivalic acid
Vzorec: C₅H₁₀O₂
MW: 102,13 g/mol
Bod varu: 164 °C (1013 hPa)
Bod tání: 32…35 °C
Hustota: 0,91 g/cm³ (20 °C)
Bod vzplanutí: 640 °C
Storage Temperature: Ambient
MDL Number: MFCD00004194
CAS číslo: 75-98-9
EINECS: 200-922-5
UN: 3261
ADR: 8,III
Merck Index: 12,07667

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR