Methyl pivalate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Methyl trimethylacetate
M-Clarity™ quality level = MQ100
M86502-100ML
M86502-500ML
SIALM86502-100MLEA
1400
CZK
SIALM86502-100ML
SIALM86502-500ML
Methyl pivalate, Sigma-Aldrich®
Methyl pivalate
Vzorec:
C₆H₁₂O₂ MW: 116,16 g/mol Bod varu: 101 °C (1013 hPa) Bod tání: < –70 °C Hustota: 0,87 g/cm³ (20 °C) Bod vzplanutí: 6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008843 CAS číslo: 598-98-1 EINECS: 209-959-1 UN: 3272 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...