Tisk…

L(-)-Glutathione (oxidised form) 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: L(-)-Glutathione, oxidized, Bi(glutathion-S-yl), GSSG, Glutathiol, Oxidized Glutathione (GSSG), Glutathione oxidized, (2S,2'S)-5,5'-(((2R,2'R)-Disulphanediylbis(1-((carboxymethyl)amino)-1-oxopropane-3,2-diyl))bis(azanediyl))bis(2-amino-5-oxopentanoic acid)

320225000 320220050
ACRO320225000EA 1850 CZK
ACRO320225000 ACRO320220050
L(-)-Glutathione (oxidised form) 98%
L(-)-Glutathione (oxidised form)
Vzorec: C₂₀H₃₂N₆O₁₂S₂
Bod tání: 178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063106
CAS číslo: 27025-41-8
EINECS: 248-170-7

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤8 % (105°C, 3 h)
HPLC ≥97.5 %
Residue after ignition ≤0.5 %
Specific optical rotation -92° to -106° (20°C, 589 nm) (c=4, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR