Pivalamide ≥98%
Dodavatel: Thermo Scientific
Synonyms:
Trimethylacetamide, 2,2,2-Trimethylacetamide
A11319.09
A11319.18
A11319.30
A11319.09EA
1160
CZK
A11319.09
A11319.18
A11319.30
Pivalamide ≥98%
Pivalamide
Purity: 98+%
CAS No.: 754-10-9
Molecular Formula: C5H11NO
Formula Weight: 101.15
Melting Point: 153-157°
Boiling Point: 212°
MDL No.: MFCD00008011
Vzorec:
C₅H₁₁NO MW: 101,15 g/mol Bod varu: 212 °C Bod tání: 153…157 °C Hustota: 1,02 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008011 CAS číslo: 754-10-9 EINECS: 212-043-4 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...