Pivalic anhydride ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Trimethylacetic anhydride, 2,2-Dimethylpropionic anhydride
B22983.36
B22983.14
B22983.22
B22983.36EA
18090
CZK
B22983.36
B22983.14
B22983.22
Pivalic anhydride ≥99%
Pivalic anhydride
Purity: 99%
CAS No.: 1538-75-6
Molecular Formula: C10H18O3
Formula Weight: 186.25
Flash Point: 63°(145°F)
Refractive Index: 1.4090
Odour: Unpleasant
Boiling Point: 193°
Beilstein Reference: 386552
UN No.: 3265
MDL No.: MFCD00008842
Vzorec:
C₁₀H₁₈O₃ MW: 186,25 g/mol Bod varu: 193 °C Hustota: 0,918 Bod vzplanutí: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008842 CAS číslo: 1538-75-6 EINECS: 216-263-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...