Tisk…

Pivaloyl chloride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Trimethylacetyl chloride

A15051.AE A15051.0F A15051.AP
A15051.AEEA 920 CZK
A15051.AE A15051.0F A15051.AP
Pivaloyl chloride ≥98%
Pivaloyl chloride

Purity: 98+% CAS No.: 3282-30-2 Molecular Formula: (CH3)3CCOCl Formula Weight: 120.58 Flash Point: 8°(46°F) Refractive Index: 1.4120 Odour: Pungent Melting Point: -57° Boiling Point: 105-106° Beilstein Reference: 385668 UN No.: 2438 MDL No.: MFCD00000709

Vzorec: C₅H₉ClO
Bod varu: 105…106 °C
Bod tání: –57 °C
Hustota: 0,985
Bod vzplanutí: 8 °C (46 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000709
CAS číslo: 3282-30-2
EINECS: 221-921-6
UN: 2438
ADR: 6.1,I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR