4,4-Dimethyl-2-pentanone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4,4-Dimethylpentan-2-one
M-Clarity™ quality level = MQ100
136875-25G
136875-5G
SIAL136875-25GEA
13710
CZK
SIAL136875-25G
SIAL136875-5G
4,4-Dimethyl-2-pentanone, Sigma-Aldrich®
4,4-Dimethyl-2-pentanone
Vzorec:
C₇H₁₄O Bod varu: 126 °C (1013 hPa) Bod tání: –64 °C Hustota: 0,808 g/cm³ (20 °C) |
MDL Number:
MFCD00008854 CAS číslo: 590-50-1 EINECS: 209-685-2 UN: 1224 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...