3,3-Dimethylbutanone ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1,1,1-Trimethylacetone, Methyl tert-butyl ketone, Pinacolone, tert-Butyl methyl ketone, 3,3-Dimethyl-2-butanone
A10289.22
A10289.36
A10289.22EA
3130
CZK
A10289.22
A10289.36
3,3-Dimethylbutanone ≥97%
3,3-Dimethylbutanone
Purity: 97%
CAS No.: 75-97-8
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 5°(41°F)
Refractive Index: 1.3970
Melting Point: -53°
Boiling Point: 105-106°
Merck Reference: 14,7440
UN No.: 1224
MDL No.: MFCD00008846
Vzorec:
C₆H₁₂O MW: 100,16 g/mol Bod varu: 105…106 °C Bod tání: –53 °C Hustota: 0,803 Bod vzplanutí: 5 °C (41 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008846 CAS číslo: 75-97-8 EINECS: 200-920-4 UN: 1224 ADR: 3,II Merck Index: 14,7440 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...