Chloromethyl pivalate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Chloromethyl trimethylacetate, Trimethylacetic acid chloromethyl ester, 2,2-Dimethylpropionic acid chloromethyl ester, Pivalic acid chloromethyl ester
M-Clarity™ quality level = MQ200
141186-100G
141186-25G
SIAL141186-100GEA
2690
CZK
SIAL141186-100G
SIAL141186-25G
Chloromethyl pivalate, Sigma-Aldrich®
Chloromethyl pivalate
Vzorec:
C₆H₁₁ClO₂ MW: 150,6 g/mol Bod varu: 70 °C (1013 hPa) Hustota: 1,0387 g/cm³ (20 °C) |
MDL Number:
MFCD00000884 CAS číslo: 18997-19-8 EINECS: 242-735-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...