1-Chloro-2,2-dimethylpropane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Chloro-1-diméthyl-2,2-propane, neo-Pentylchloride
M-Clarity™ quality level = MQ100
274496-25G
274496-5G
SIAL274496-25GEA
6210
CZK
SIAL274496-25G
SIAL274496-5G
1-Chloro-2,2-dimethylpropane, Sigma-Aldrich®
1-Chloro-2,2-dimethylpropane
Vzorec:
C₅H₁₁Cl MW: 106,6 g/mol Bod varu: 84 °C (1013 hPa) Bod tání: –20 °C Hustota: 0,86 g/cm³ (20 °C) |
MDL Number:
MFCD00000941 CAS číslo: 753-89-9 EINECS: 212-040-8 UN: 1107 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...