Isovaleraldehyde, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Methylbutyraldehyde
MQ300
W269204-1KG-K
W269204-20KG-K
W269204-4KG-K
SIALW269204-1KG-KEA
4010
CZK
SIALW269204-1KG-K
SIALW269204-20KG-K
SIALW269204-4KG-K
Isovaleraldehyde, Sigma-Aldrich®
Isovaleraldehyde
Vzorec:
C₅H₁₀O MW: 86.13 Bod varu: 90 Bod tání: –60 °C Hustota: 0.8 g/cm³ (20 °C) Bod vzplanutí: 0 Storage Temperature: Refrigerator |
MDL Number:
MFCD00007014 CAS číslo: 590-86-3 EINECS: 209-691-5 UN: 2058 ADR: 3 II Merck Index: 13,05250 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...