Tisk…
Safety shoes, lace-up, Basic, Lenny S3, Jori
Dodavatel: ELTEN
12321/36 12321/37 12321/38 12321/39 12321/40 12321/41 12321/42 12321/43 12321/44 12321/45 12321/46 12321/47 12321/48
ELTN12321/36EA 1230 CZK
ELTN12321/36 ELTN12321/37 ELTN12321/38 ELTN12321/39 ELTN12321/40 ELTN12321/41 ELTN12321/42 ELTN12321/43 ELTN12321/44 ELTN12321/45 ELTN12321/46 ELTN12321/47 ELTN12321/48
Safety shoes, lace-up, Basic, Lenny S3, Jori
Obuv
Durable safety shoes made from wear resistant coated leather, skin friendly textile lining, a full padded bellows tongue and collar padding, plus a dual-density PU/PU New Basic sole (oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole
  • Anti-static soft fleece insole
  • PU scuff cap protects the upper material
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, or assembly and whereMore Product Information