n-Heneicosane, Sigma-Aldrich®
Dodavatel: Merck
MQ100
286052-10G
286052-1G
SIAL286052-10GEA
4720
CZK
SIAL286052-10G
SIAL286052-1G
n-Heneicosane, Sigma-Aldrich®
n-Heneicosane
Vzorec:
H₃C(CH₂)₁₉CH₃ MW: 296.58 Bod varu: 356 Bod tání: 40…41 °C Hustota: 0.7919 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009346 CAS číslo: 629-94-7 EINECS: 211-118-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...