Tisk…

n-Dotriacontane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dotriacontane

M-Clarity™ quality level = MQ200

D223107-25G D223107-5G
SIALD223107-25GEA 4410 CZK
SIALD223107-25G SIALD223107-5G
n-Dotriacontane, Sigma-Aldrich®
n-Dotriacontane
Vzorec: H₃C(CH₂)₃₀CH₃
Bod varu: 467 °C (1013 hPa)
Hustota: 0,7798 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009411
CAS číslo: 544-85-4
EINECS: 208-881-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR