Tisk…

Sodium methanesulphonate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

304506-100G 304506-500G 304506-5G
SIAL304506-100GEA 3110 CZK
SIAL304506-100G SIAL304506-500G SIAL304506-5G
Sodium methanesulphonate, Sigma-Aldrich®
Sodium methanesulphonate
Vzorec: CH₃NaO₃S
MDL Number: MFCD00064389
CAS číslo: 2386-57-4
EINECS: 219-203-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR