Tisk…

Kyselina 1-dodekansulfonová sodná sůl ≥99%

Dodavatel: Thermo Scientific

Synonyms: Sodium 1-dodecanesulphonate, Dodecane-1-sulphonic acid sodium salt, Sodium 1-dodecansulphonate, 1-Dodecanesulphonic acid, sodium salt, Sodium dodecane-1-sulphonate

A12829.03 A12829.14 A12829.09
A12829.03EA 1210 CZK
A12829.03 A12829.14 A12829.09
Kyselina 1-dodekansulfonová sodná sůl ≥99%
Kyselina 1-dodekansulfonová sodná sůl

Purity: 99% CAS No.: 2386-53-0 Molecular Formula: CH3(CH2)11SO3Na Formula Weight: 272.39 Melting Point: 300° Beilstein Reference: 3919536

Vzorec: C₁₂H₂₅NaO₃S
MW: 272,38 g/mol
Bod tání: >300 °C
Hustota: 1,218 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007527
CAS číslo: 2386-53-0
EINECS: 219-200-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR