Tisk…

Ethanesulphonic acid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

186260-25G 186260-5G
SIAL186260-25GEA 2550 CZK
SIAL186260-25G SIAL186260-5G
Ethanesulphonic acid, Sigma-Aldrich®
Ethanesulphonic acid
Vzorec: C₂H₆O₃S
MW: 110.13
Bod varu: 123 °C (1 mmHg)
Bod tání: –17 °C
Hustota: 1.343 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007529
CAS číslo: 594-45-6
EINECS: 209-843-0
UN: 2586
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR