Tisk…

Ethanesulphonic acid ≥96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

14891.09 14891.18 14891.30
14891.09EA 1460 CZK
14891.09 14891.18 14891.30
Ethanesulphonic acid ≥96%
Ethanesulphonic acid

Purity: 96% CAS No.: 594-45-6 Molecular Formula: CH3CH2SO3H Formula Weight: 110.13 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.343 Refractive Index: 1.4350 Appearance: Colourless to yellow Melting Point: -17° Boiling Point: 122-123°/0.01mm Beilstein Reference: 1743071 UN No.: 2586 MDL No.: MFCD00007529

Vzorec: C₂H₆O₃S
MW: 110.13
Bod varu: 122…123 °C (0.01 mmHg)
Bod tání: –17 °C
Hustota: 1.343
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007529
CAS číslo: 594-45-6
EINECS: 209-843-0
UN: 2586
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR