Ethyl methanesulphonate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
M0880-1G
M0880-100G
M0880-10G
M0880-25G
M0880-5G
SIALM0880-1GEA
1390
CZK
SIALM0880-1G
SIALM0880-100G
SIALM0880-10G
SIALM0880-25G
SIALM0880-5G
Ethyl methanesulphonate, Sigma-Aldrich®
Ethyl methanesulphonate
Vzorec:
C₃H₈O₃S Bod varu: 213 °C (1013 hPa) Hustota: 1,2079 g/cm³ (20 °C) Bod vzplanutí: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007559 CAS číslo: 62-50-0 EINECS: 200-536-7 UN: 2810 ADR: 6.1,II Merck Index: 13,03859 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...