Ethanesulphonyl chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
471542-100ML
471542-500ML
SIAL471542-100MLEA
2280
CZK
SIAL471542-100ML
SIAL471542-500ML
Ethanesulphonyl chloride, Sigma-Aldrich®
Ethanesulphonyl chloride
Vzorec:
C₂H₅ClO₂S MW: 128,58 g/mol Bod varu: 177 °C (1013 hPa) Bod tání: –68 °C Hustota: 1,36 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007460 CAS číslo: 594-44-5 EINECS: 209-842-5 UN: 2927 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...