1-Pentanethiol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
P7908-100G-A
P7908-25G-A
SIALP7908-100G-AEA
11020
CZK
SIALP7908-100G-A
SIALP7908-25G-A
1-Pentanethiol, Sigma-Aldrich®
1-Pentanethiol
Vzorec:
C₅H₁₂S Bod varu: 127 °C (1013 hPa) Bod tání: –76 °C Hustota: 0,841 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004907 CAS číslo: 110-66-7 EINECS: 203-789-1 UN: 1111 ADR: 3,II Merck Index: 12,00653 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...