Tisk…

1-Octadecanethiol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: n-Octadecyl mercaptan, n-Octadecanethiol, Stearyl mercaptane, 1-Mercaptooctadecane, Octadecane-1-thiol

M-Clarity™ quality level = MQ200

O1858-100ML O1858-25ML
SIALO1858-100MLEA 4660 CZK
SIALO1858-100ML SIALO1858-25ML
1-Octadecanethiol, Sigma-Aldrich®
1-Octadecanethiol
Vzorec: C₁₈H₃₈S
MW: 286,57 g/mol
Bod varu: 204 °C (1013 hPa)
Bod tání: 28…31 °C
Hustota: 0,8475 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004886
CAS číslo: 2885-00-9
EINECS: 220-744-1
UN: 3335
ADR: 9,

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR