Ethanethiol, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Ethyl mercaptan, 1-Mercaptoethane
M-Clarity™ quality level = MQ200
E3708-100ML
E3708-500ML
SIALE3708-100MLEA
1230
CZK
SIALE3708-100ML
SIALE3708-500ML
Ethanethiol, Sigma-Aldrich®
Ethanethiol
Vzorec:
C₂H₆S MW: 62,14 g/mol Bod varu: 35 °C (1013 hPa) Bod tání: –148 °C Hustota: 0,846 g/cm³ (20 °C) Bod vzplanutí: –45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004887 CAS číslo: 75-08-1 EINECS: 200-837-3 UN: 2363 ADR: 3,I Merck Index: 12,03771 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...