Methyl thiocyanate ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H27879.AA
H27879.AC
H27879.AE
H27879.AAEA
827
CZK
H27879.AA
H27879.AC
H27879.AE
Methyl thiocyanate ≥99%
Methyl thiocyanate
Purity: 99%
CAS No.: 556-64-9
Molecular Formula: CH3SCN
Formula Weight: 73.11
Flash Point: 38°(100°F)
Refractive Index: 1.4680
Odour: Pungent
Melting Point: -5°
Boiling Point: 131°
UN No.: 2929
MDL No.: MFCD00001830
Vzorec:
C₂H₃NS Bod varu: 131 °C Bod tání: –5 °C Hustota: 1,068 Bod vzplanutí: 38 °C (100 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001830 CAS číslo: 556-64-9 EINECS: 209-134-6 UN: 2929 ADR: 6.1,II Merck Index: 12,06209 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...